Assay Details
| Gene Symbol: ERG28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719637.3 | XP_002719683.1 | 4 - 5 | 470 | 105 | |
| XM_008272027.2 | XP_008270249.1 | 4 - 5 | 454 | 105 | ||
| XM_008272028.2 | XP_008270250.1 | 4 - 5 | 453 | 105 | ||
| XM_017349416.1 | XP_017204905.1 | 4 - 5 | 436 | 105 | ||
More Information
Set Membership: |
|

ERG28
Genomic Map