Assay Details
| Gene Symbol: BORCS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718588.3 | XP_002718634.1 | 3 - 4 | 323 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BORCS7| Gene Symbol: BORCS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718588.3 | XP_002718634.1 | 3 - 4 | 323 | 100 | |
Set Membership: |
|