Assay Details
| Gene Symbol: PABPC1L2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350009.1 | XP_017205498.1 | 1 | 1546 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PABPC1L2B| Gene Symbol: PABPC1L2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350009.1 | XP_017205498.1 | 1 | 1546 | 74 | |
Set Membership: |
|