Assay Details
| Gene Symbol: STIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709383.3 | XP_002709429.1 | 10 - 11 | 1766 | 60 | |
| XM_008274823.2 | XP_008273045.1 | 9 - 10 | 1741 | 60 | ||
More Information
Set Membership: |
|

STIM2
Genomic Map