Assay Details
| Gene Symbol: KCTD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260850.2 | XP_008259072.1 | 3 - 4 | 1596 | 82 | |
| XM_008260851.2 | XP_008259073.1 | 2 - 3 | 194 | 82 | ||
| XM_008260852.2 | XP_008259074.1 | 2 - 3 | 1567 | 82 | ||
| XM_017343904.1 | XP_017199393.1 | 4 - 5 | 1994 | 82 | ||
| XM_017343905.1 | XP_017199394.1 | 2 - 3 | 214 | 82 | ||
| XM_017343906.1 | XP_017199395.1 | 2 - 3 | 434 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008260850.2, XM_008260851.2, XM_008260852.2, XM_017343906.1 transcripts.
Set Membership: |
|

KCTD6
Genomic Map