Assay Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714715.3 | XP_002714761.1 | 2 - 3 | 251 | 78 | |
| XM_017345356.1 | XP_017200845.1 | 2 - 3 | 259 | 78 | ||
More Information
Set Membership: |
|

HDDC2
Genomic Map