Assay Details
Gene Symbol: WASHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711226.3 | XP_002711272.1 | 1 - 2 | 153 | 78 | |
XM_008256980.2 | XP_008255202.1 | 1 - 2 | 159 | 78 | ||
XM_017342062.1 | XP_017197551.1 | 1 - 2 | 162 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
