Assay Details
| Gene Symbol: SERTAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723260.3 | XP_002723306.1 | 150 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723260.3 transcripts.
Set Membership: |
|

SERTAD1
Genomic Map