Assay Details
| Gene Symbol: GNAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713561.3 | XP_002713607.1 | 7 - 8 | 932 | 58 | |
| XM_008261023.2 | XP_008259245.1 | 7 - 8 | 1499 | 58 | ||
| XM_008261024.2 | XP_008259246.1 | 8 - 9 | 864 | 58 | ||
More Information
Set Membership: |
|

GNAL
Genomic Map