Assay Details
| Gene Symbol: PCYT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719992.3 | XP_002720038.1 | 6 - 7 | 897 | 64 | |
| XM_008272466.2 | XP_008270688.1 | 7 - 8 | 951 | 64 | ||
| XM_008272467.2 | XP_008270689.2 | 6 - 7 | 911 | 64 | ||
| XM_008272468.2 | XP_008270690.1 | 6 - 7 | 870 | 64 | ||
More Information
Set Membership: |
|

PCYT1B
Genomic Map