Assay Details
| Gene Symbol: MAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263749.1 | XP_008261971.1 | 2 - 3 | 592 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008263749.1 transcripts.
Set Membership: |
|

MAS1
Genomic Map