Assay Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723894.3 | XP_002723940.1 | 5 - 6 | 797 | 60 | |
| XM_008252766.2 | XP_008250988.1 | 5 - 6 | 930 | 60 | ||
| XM_017340013.1 | XP_017195502.1 | 5 - 6 | 648 | 60 | ||
More Information
Set Membership: |
|

SPOUT1
Genomic Map