Assay Details
| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350870.1 | XP_017206359.1 | 760 | 78 | ||
| XM_017350871.1 | XP_017206360.1 | 760 | 78 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRA3| Gene Symbol: GABRA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350870.1 | XP_017206359.1 | 760 | 78 | ||
| XM_017350871.1 | XP_017206360.1 | 760 | 78 | |||
Set Membership: |
|