Assay Details
| Gene Symbol: CARMIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348213.1 | XP_017203702.1 | 10 - 11 | 946 | 56 | |
| XM_017348214.1 | XP_017203703.1 | 10 - 11 | 946 | 56 | ||
More Information
Set Membership: |
|

CARMIL3
Genomic Map