Assay Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265129.2 | XP_008263351.1 | 10 - 11 | 1205 | 68 | |
| XM_008265130.2 | XP_008263352.1 | 11 - 12 | 1346 | 68 | ||
| XM_008265131.1 | XP_008263353.1 | 10 - 11 | 1132 | 68 | ||
| XM_008265132.2 | XP_008263354.1 | 9 - 10 | 1116 | 68 | ||
| XM_017346231.1 | XP_017201720.1 | 10 - 11 | 1114 | 68 | ||
More Information
Set Membership: |
|

ALG6
Genomic Map