Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341249.1 | XP_017196738.1 | 1 - 2 | 2232 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

INSYN2B| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017341249.1 | XP_017196738.1 | 1 - 2 | 2232 | 57 | |
Set Membership: |
|