Assay Details
| Gene Symbol: LOC100341431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261630.2 | XP_008259852.1 | 18 - 19 | 1936 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100341431| Gene Symbol: LOC100341431 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261630.2 | XP_008259852.1 | 18 - 19 | 1936 | 122 | |
Set Membership: |
|