Assay Details
| Gene Symbol: LOC100343179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250636.2 | XP_008248858.1 | 1425 | 115 | ||
| XM_008250637.2 | XP_008248859.1 | 1259 | 115 | |||
| XM_008250638.2 | XP_008248860.1 | 1140 | 115 | |||
More Information
Set Membership: |
|

LOC100343179
Genomic Map