Assay Details
Gene Symbol: ZNF346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710418.3 | XP_002710464.1 | 1 - 2 | 224 | 78 | |
XM_008255454.2 | XP_008253676.1 | 1 - 2 | 225 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
