Assay Details
| Gene Symbol: TOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274459.1 | XP_008272681.1 | 436 | 64 | ||
| XM_008274460.1 | XP_008272682.1 | 436 | 64 | |||
| XM_008274462.1 | XP_008272684.1 | 421 | 64 | |||
| XM_008274463.2 | XP_008272685.1 | 600 | 64 | |||
| XM_017350598.1 | XP_017206087.1 | 436 | 64 | |||
More Information
Set Membership: |
|

TOX2
Genomic Map