Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258376.1 | XP_008256598.1 | 5 | 558 | 70 | |
| XM_008258377.2 | XP_008256599.1 | 5 | 521 | 70 | ||
| XM_008258378.2 | XP_008256600.1 | 5 | 521 | 70 | ||
| XM_008258379.1 | XP_008256601.1 | 5 | 568 | 70 | ||
More Information
Set Membership: |
|

IMMP2L
Genomic Map