Assay Details
| Gene Symbol: SPART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720586.3 | XP_002720632.1 | 1511 | 102 | ||
| XM_008273553.2 | XP_008271775.1 | 1279 | 102 | |||
| XM_008273554.2 | XP_008271776.1 | 1911 | 102 | |||
More Information
Set Membership: |
|

SPART
Genomic Map