Assay Details
| Gene Symbol: STIMATE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713252.3 | XP_002713298.1 | 7 - 8 | 849 | 102 | |
| XM_008260802.2 | XP_008259024.1 | 7 - 8 | 849 | 102 | ||
More Information
Set Membership: |
|

STIMATE
Genomic Map