Assay Details
| Gene Symbol: TRIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017350274.1 | XP_017205763.1 | 255 | 70 | ||
| XM_017350275.1 | XP_017205764.1 | 239 | 70 | |||
| XM_017350276.1 | XP_017205765.1 | 295 | 70 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017350274.1, XM_017350275.1, XM_017350276.1 transcripts.
Set Membership: |
|

TRIM13
Genomic Map