Assay Details
| Gene Symbol: SUV39H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717451.3 | XP_002717497.1 | 5 - 6 | 1207 | 79 | |
| XM_008268314.2 | XP_008266536.1 | 5 - 6 | 1245 | 79 | ||
| XM_008268316.2 | XP_008266538.1 | 4 - 5 | 1114 | 79 | ||
| XM_017347757.1 | XP_017203246.1 | 4 - 5 | 1087 | 79 | ||
More Information
Set Membership: |
|

SUV39H2
Genomic Map