Assay Details
| Gene Symbol: DBF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271629.2 | XP_008269851.1 | 10 - 11 | 1008 | 87 | |
| XM_008271630.2 | XP_008269852.1 | 10 - 11 | 901 | 87 | ||
| XM_008271632.2 | XP_008269854.1 | 9 - 10 | 804 | 87 | ||
More Information
Set Membership: |
|

DBF4B
Genomic Map