Assay Details
| Gene Symbol: LOC100347090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720525.2 | XP_002720571.1 | 1092 | 106 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100347090| Gene Symbol: LOC100347090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720525.2 | XP_002720571.1 | 1092 | 106 | ||
Set Membership: |
|