Assay Details
| Gene Symbol: LOC100358417 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008254993.2 | XP_008253215.2 | 1029 | 58 | ||
| XM_017341035.1 | XP_017196524.1 | 1029 | 58 | |||
More Information
Set Membership: |
|

LOC100358417
Genomic Map