Assay Details
| Gene Symbol: RAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714103.3 | XP_002714149.1 | 6 | 1926 | 93 | |
| XM_008262104.2 | XP_008260326.1 | 6 | 2068 | 93 | ||
More Information
Set Membership: |
|

RAD1
Genomic Map