Assay Details
| Gene Symbol: KCTD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270790.2 | XP_008269012.1 | 1 | 481 | 68 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008270790.2 transcripts.
Set Membership: |
|

KCTD11
Genomic Map