Assay Details
| Gene Symbol: MAPK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722652.3 | XP_002722698.1 | 565 | 56 | ||
| XM_008250339.2 | XP_008248561.1 | 981 | 56 | |||
| XM_008250340.2 | XP_008248562.1 | 671 | 56 | |||
| XM_017338597.1 | XP_017194086.1 | 939 | 56 | |||
More Information
Set Membership: |
|

MAPK7
Genomic Map