Assay Details
| Gene Symbol: PROK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715430.3 | XP_002715476.1 | 2 - 3 | 326 | 90 | |
| XM_017345608.1 | XP_017201097.1 | 2 - 3 | 342 | 90 | ||
More Information
Set Membership: |
|

PROK1
Genomic Map