Assay Details
| Gene Symbol: ZSCAN30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261199.2 | XP_008259421.1 | 2 - 3 | 664 | 63 | |
| XM_008261200.2 | XP_008259422.1 | 4 - 5 | 941 | 63 | ||
| XM_017344105.1 | XP_017199594.1 | 4 - 5 | 941 | 63 | ||
| XM_017344106.1 | XP_017199595.1 | 3 - 4 | 865 | 63 | ||
| XM_017344107.1 | XP_017199596.1 | 3 - 4 | 746 | 63 | ||
| XM_017344108.1 | XP_017199597.1 | 4 - 5 | 940 | 63 | ||
More Information
Set Membership: |
|

ZSCAN30
Genomic Map