Assay Details
| Gene Symbol: IL1RAPL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272830.2 | XP_008271052.1 | 8 - 9 | 2033 | 62 | |
| XM_008272833.2 | XP_008271055.1 | 8 - 9 | 2033 | 62 | ||
More Information
Set Membership: |
|

IL1RAPL2
Genomic Map