Assay Details
| Gene Symbol: PRSS53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721732.3 | XP_002721778.2 | 857 | 60 | ||
| XM_017337748.1 | XP_017193237.1 | 859 | 60 | |||
| XM_017337749.1 | XP_017193238.1 | 858 | 60 | |||
More Information
Set Membership: |
|

PRSS53
Genomic Map