Assay Details
| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714555.3 | XP_002714601.1 | 2 - 3 | 310 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRAP2| Gene Symbol: MRAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714555.3 | XP_002714601.1 | 2 - 3 | 310 | 73 | |
Set Membership: |
|