Assay Details
| Gene Symbol: C13H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715464.3 | XP_002715510.2 | 1 - 2 | 188 | 74 | |
| XM_008264382.2 | XP_008262604.1 | 1 - 2 | 190 | 74 | ||
More Information
Set Membership: |
|

C13H1orf189
Genomic Map