Assay Details
| Gene Symbol: C13H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715464.3 | XP_002715510.2 | 2 - 3 | 237 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C13H1orf189| Gene Symbol: C13H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715464.3 | XP_002715510.2 | 2 - 3 | 237 | 106 | |
Set Membership: |
|