Assay Details
| Gene Symbol: C13H1orf189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715464.3 | XP_002715510.2 | 3 - 4 | 364 | 51 | |
| XM_008264382.2 | XP_008262604.1 | 3 - 4 | 390 | 51 | ||
More Information
Set Membership: |
|

C13H1orf189
Genomic Map