Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710842.3 | XP_002710888.1 | 8 - 9 | 1230 | 62 | |
| XM_017341765.1 | XP_017197254.1 | 6 - 7 | 867 | 62 | ||
More Information
Set Membership: |
|

TRMT6
Genomic Map