Assay Details
| Gene Symbol: TBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714832.3 | XP_002714878.1 | 5 - 6 | 544 | 71 | |
| XM_008263539.2 | XP_008261761.1 | 4 - 5 | 703 | 71 | ||
| XM_008263540.2 | XP_008261762.1 | 4 - 5 | 470 | 71 | ||
| XM_008263541.2 | XP_008261763.1 | 4 - 5 | 406 | 71 | ||
| XM_017345381.1 | XP_017200870.1 | 4 - 5 | 441 | 71 | ||
More Information
Set Membership: |
|

TBPL1
Genomic Map