Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275053.2 | XP_008273275.1 | 395 | 121 | ||
| XM_008275054.2 | XP_008273276.1 | 396 | 121 | |||
| XM_008275056.2 | XP_008273278.1 | 407 | 121 | |||
More Information
Set Membership: |
|

TENT4B
Genomic Map