Assay Details
| Gene Symbol: TENT4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275053.2 | XP_008273275.1 | 1082 | 102 | ||
| XM_008275054.2 | XP_008273276.1 | 1083 | 102 | |||
More Information
Set Membership: |
|

TENT4B
Genomic Map