Assay Details
| Gene Symbol: C2CD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258970.2 | XP_008257192.1 | 4 - 5 | 595 | 119 | |
| XM_017343030.1 | XP_017198519.1 | 4 - 5 | 595 | 119 | ||
More Information
Set Membership: |
|

C2CD6
Genomic Map