Assay Details
| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248414.1 | XP_008246636.1 | 792 | 62 | ||
| XM_008248415.2 | XP_008246637.1 | 445 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUN3| Gene Symbol: SUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248414.1 | XP_008246636.1 | 792 | 62 | ||
| XM_008248415.2 | XP_008246637.1 | 445 | 62 | |||
Set Membership: |
|