Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261151.1 | XP_008259373.1 | 5 - 6 | 1279 | 84 | |
| XM_017344087.1 | XP_017199576.1 | 4 - 5 | 1064 | 84 | ||
More Information
Set Membership: |
|

CABYR
Genomic Map