Assay Details
| Gene Symbol: CCDC157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721186.3 | XP_002721232.1 | 1816 | 58 | ||
| XM_008274707.2 | XP_008272929.1 | 1949 | 58 | |||
| XM_008274708.2 | XP_008272930.1 | 1872 | 58 | |||
More Information
Set Membership: |
|

CCDC157
Genomic Map