Assay Details
| Gene Symbol: C1H11orf42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708754.3 | XP_002708800.1 | 1 - 2 | 2267 | 86 | |
| XM_008265186.2 | XP_008263408.1 | 1 - 2 | 2267 | 86 | ||
More Information
Set Membership: |
|

C1H11orf42
Genomic Map