Assay Details
| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271247.1 | XP_008269469.1 | 2 - 3 | 327 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DYNLL2| Gene Symbol: DYNLL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271247.1 | XP_008269469.1 | 2 - 3 | 327 | 93 | |
Set Membership: |
|