Assay Details
| Gene Symbol: LOC100358830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338435.1 | XP_017193924.1 | 828 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017338435.1 transcripts.
Set Membership: |
|

LOC100358830
Genomic Map